missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-4-hydroxybenzonitrile, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2610703

 View more versions of this product

Catalog No. AAH2610703


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoro-4-hydroxybenzonitrile
133°C to 135°C
MFCD00215834
UN3439
DPSSSDFTLVUJDH-UHFFFAOYSA-N
3-fluoro-4-hydroxybenzonitrile
2782944
98%
405-04-9
C7H4FNO
1 g
4-cyano-2-fluorophenol, 2-fluoro-4-cyanophenol, benzonitrile, 3-fluoro-4-hydroxy, 3-fluoro-4-hydroxy-benzonitril, 3-fluoro-4-hydroxy benzonitrile, 3-fluoro-4-hydroxy-benzonitrile, 3-fluoro-4-hydroxybenzenecarbonitrile, acmc-209jep, 4-cyano-2-fluoro phenol, ksc493s8d
C1=CC(=C(C=C1C#N)F)O
137.113
137.11

Chemical Identifiers

405-04-9
137.113
DPSSSDFTLVUJDH-UHFFFAOYSA-N
2782944
C1=CC(=C(C=C1C#N)F)O
C7H4FNO
MFCD00215834
4-cyano-2-fluorophenol, 2-fluoro-4-cyanophenol, benzonitrile, 3-fluoro-4-hydroxy, 3-fluoro-4-hydroxy-benzonitril, 3-fluoro-4-hydroxy benzonitrile, 3-fluoro-4-hydroxy-benzonitrile, 3-fluoro-4-hydroxybenzenecarbonitrile, acmc-209jep, 4-cyano-2-fluoro phenol, ksc493s8d
3-fluoro-4-hydroxybenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only