Learn More
3-Fluoro-4-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1951603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Fluoro-4-methylbenzenesulfonyl chloride | |
46°C to 50°C | |
C7H6ClFO2S | |
1 g | |
Moisture sensitive | |
YDUHIMCRFRIVFI-UHFFFAOYSA-N | |
3-fluoro-4-methylbenzenesulfonyl chloride | |
2778600 | |
97% |
90260-13-2 | |
>110°C (230°F) | |
MFCD03094389 | |
UN3261 | |
3-fluoro-4-methylbenzene-1-sulfonyl chloride, 3-fluoro-4-methylbenzenesulfonylchloride, 3-fluoro-4-methylbenzenesulphonyl chloride, 3-fluoro-4-methyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-methyl, 3-fluoro-p-toluenesulphonyl chloride, 3-fluoro-p-toluenesulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-methyl-9ci, chloro 3-fluoro-4-methylphenyl sulfone, pubchem11718 | |
CC1=C(C=C(C=C1)S(=O)(=O)Cl)F | |
208.631 | |
208.64 |
Chemical Identifiers
90260-13-2 | |
208.631 | |
YDUHIMCRFRIVFI-UHFFFAOYSA-N | |
2778600 | |
CC1=C(C=C(C=C1)S(=O)(=O)Cl)F |
C7H6ClFO2S | |
MFCD03094389 | |
3-fluoro-4-methylbenzene-1-sulfonyl chloride, 3-fluoro-4-methylbenzenesulfonylchloride, 3-fluoro-4-methylbenzenesulphonyl chloride, 3-fluoro-4-methyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-methyl, 3-fluoro-p-toluenesulphonyl chloride, 3-fluoro-p-toluenesulfonyl chloride, benzenesulfonyl chloride, 3-fluoro-4-methyl-9ci, chloro 3-fluoro-4-methylphenyl sulfone, pubchem11718 | |
3-fluoro-4-methylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only