missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-nitroaniline 95.0+%, TCI America™

$70.84 - $281.48

Chemical Identifiers

CAS 2369-13-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.116
MDL Number MFCD03412239
InChI Key KKQPNAPYVIIXFB-UHFFFAOYSA-N
Synonym 4-amino-2-fluoronitrobenzene, 3-fluoro-4-nitro-phenylamine, 3-fluoro-4-nitro-1-benzenamine, benzenamine, 3-fluoro-4-nitro, fluoronitroaniline2, pubchem4474, 3-fluoro-4-nitroanil, 3-fluor-4-nitroaniline, 4-nitro-3-fluoroanilin, acmc-1cirv
PubChem CID 75401
IUPAC Name 3-fluoro-4-nitroaniline
SMILES C1=CC(=C(C=C1N)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F07871G
View Documents
TCI America
F07871G
1 g
Each for $70.84
Only null left
 
F07875G
View Documents
TCI America
F07875G
5 g
Each for $281.48
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2369-13-3
156.116
KKQPNAPYVIIXFB-UHFFFAOYSA-N
75401
C1=CC(=C(C=C1N)F)[N+](=O)[O-]
C6H5FN2O2
MFCD03412239
4-amino-2-fluoronitrobenzene, 3-fluoro-4-nitro-phenylamine, 3-fluoro-4-nitro-1-benzenamine, benzenamine, 3-fluoro-4-nitro, fluoronitroaniline2, pubchem4474, 3-fluoro-4-nitroanil, 3-fluor-4-nitroaniline, 4-nitro-3-fluoroanilin, acmc-1cirv
3-fluoro-4-nitroaniline

Specifications

2369-13-3
C6H5FN2O2
1 g
KKQPNAPYVIIXFB-UHFFFAOYSA-N
3-fluoro-4-nitroaniline
75401
≥95.0% (GC)
3-Fluoro-4-nitroaniline
161°C
MFCD03412239
4-amino-2-fluoronitrobenzene, 3-fluoro-4-nitro-phenylamine, 3-fluoro-4-nitro-1-benzenamine, benzenamine, 3-fluoro-4-nitro, fluoronitroaniline2, pubchem4474, 3-fluoro-4-nitroanil, 3-fluor-4-nitroaniline, 4-nitro-3-fluoroanilin, acmc-1cirv
C1=CC(=C(C=C1N)F)[N+](=O)[O-]
156.116
156.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'