missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Fluoro-5-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Chemical Identifiers
Synonym | 3-Fluoro-5-(4-morpholinylmethyl)phenylboronic acid pinacol ester |
---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH62773MD
|
Thermo Scientific Chemicals
H62773MD |
250mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3-Fluoro-5-(4-morpholinylmethyl)phenylboronic acid pinacol ester |
Specifications
250mg | |
321.19 | |
3-Fluoro-5-(4-morpholinylmethyl)benzeneboronic acid pinacol ester |
3-Fluoro-5-(4-morpholinylmethyl)phenylboronic acid pinacol ester | |
96% |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures