missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-5-iodoaniline, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3445706

 View more versions of this product

Catalog No. AAH3445706


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoro-5-iodoaniline
C6H5FIN
5 g
5-fluoro-3-iodoaniline, 3-fluoro-5-iodo-phenylamine, pubchem4458, 3-fluoro-5-iodo aniline, 3-fluoro-5-iodophenylamine, 5-fluoro-3-iodophenylamine, benzenamine,3-fluoro-5-iodo, benzenamine, 3-fluoro-5-iodo
C1=C(C=C(C=C1F)I)N
237.016
237.02
660-49-1
MFCD04116466
Light sensitive
HTEPMXLKKDKDAQ-UHFFFAOYSA-N
3-fluoro-5-iodoaniline
2758986
96%

Chemical Identifiers

660-49-1
237.016
HTEPMXLKKDKDAQ-UHFFFAOYSA-N
2758986
C1=C(C=C(C=C1F)I)N
C6H5FIN
MFCD04116466
5-fluoro-3-iodoaniline, 3-fluoro-5-iodo-phenylamine, pubchem4458, 3-fluoro-5-iodo aniline, 3-fluoro-5-iodophenylamine, 5-fluoro-3-iodophenylamine, benzenamine,3-fluoro-5-iodo, benzenamine, 3-fluoro-5-iodo
3-fluoro-5-iodoaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only