missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-5-nitrotoluene, 98%, Thermo Scientific™

$132.95 - $520.81

Chemical Identifiers

CAS 499-08-1
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD06797943
InChI Key IPMURRZVEWZEPP-UHFFFAOYSA-N
Synonym 3-fluoro-5-nitrotoluene, benzene, 1-fluoro-3-methyl-5-nitro, 3-fluoro-5-methylnitrobenzene, pubchem1625, acmc-209kix, ksc494a3n, 3-fluoro-5-methyl-nitrobenzene, 5-fluoro-3-nitrotoluene, 1-fluoro-3-methyl-5-nitro-benzene, 1-fluoranyl-3-methyl-5-nitro-benzene
PubChem CID 15065244
IUPAC Name 1-fluoro-3-methyl-5-nitrobenzene
SMILES CC1=CC(=CC(=C1)F)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6110003
View Documents
Thermo Scientific Chemicals
H6110003
1 g
Each for $132.95
 
AAH6110006
View Documents
Thermo Scientific Chemicals
H6110006
5 g
Each for $520.81
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

499-08-1
155.128
IPMURRZVEWZEPP-UHFFFAOYSA-N
15065244
CC1=CC(=CC(=C1)F)[N+](=O)[O-]
C7H6FNO2
MFCD06797943
3-fluoro-5-nitrotoluene, benzene, 1-fluoro-3-methyl-5-nitro, 3-fluoro-5-methylnitrobenzene, pubchem1625, acmc-209kix, ksc494a3n, 3-fluoro-5-methyl-nitrobenzene, 5-fluoro-3-nitrotoluene, 1-fluoro-3-methyl-5-nitro-benzene, 1-fluoranyl-3-methyl-5-nitro-benzene
1-fluoro-3-methyl-5-nitrobenzene

Specifications

499-08-1
C7H6FNO2
1 g
IPMURRZVEWZEPP-UHFFFAOYSA-N
1-fluoro-3-methyl-5-nitrobenzene
15065244
98%
3-Fluoro-5-nitrotoluene
Yellow
MFCD06797943
3-fluoro-5-nitrotoluene, benzene, 1-fluoro-3-methyl-5-nitro, 3-fluoro-5-methylnitrobenzene, pubchem1625, acmc-209kix, ksc494a3n, 3-fluoro-5-methyl-nitrobenzene, 5-fluoro-3-nitrotoluene, 1-fluoro-3-methyl-5-nitro-benzene, 1-fluoranyl-3-methyl-5-nitro-benzene
CC1=CC(=CC(=C1)F)[N+](=O)[O-]
155.128
155.13
Crystalline Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'