missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoroanisole, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1093130

 View more versions of this product

Catalog No. AAA1093130


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoroanisole
-35°C
158°C to 159°C
C7H7FO
MFCD00000335
UN1993
3-fluoroanisole, m-fluoroanisole, benzene, 1-fluoro-3-methoxy, 1-fluoro-3-methoxy-benzene, 2-fluoro-4-methoxybenzene, fr co1 wln, 3-fluoroanisol, 3-fluoro-anisole, 3-fluoro anisole, anisole, m-fluoro
COC1=CC=CC(F)=C1
126.13
126.13
456-49-5
1.104
43°C (109°F)
1.489
250 g
1858895
MFJNOXOAIFNSBX-UHFFFAOYSA-N
1-fluoro-3-methoxybenzene
9975
99%

Chemical Identifiers

456-49-5
126.13
MFJNOXOAIFNSBX-UHFFFAOYSA-N
9975
COC1=CC=CC(F)=C1
C7H7FO
MFCD00000335
3-fluoroanisole, m-fluoroanisole, benzene, 1-fluoro-3-methoxy, 1-fluoro-3-methoxy-benzene, 2-fluoro-4-methoxybenzene, fr co1 wln, 3-fluoroanisol, 3-fluoro-anisole, 3-fluoro anisole, anisole, m-fluoro
1-fluoro-3-methoxybenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 207-267-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only