missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Fluoroazetidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America F11861G
Specifications
3-Fluoroazetidine Hydrochloride | |
130°C | |
C3H7ClFN | |
1 g | |
PXFUWRWCKSLCLS-UHFFFAOYSA-N | |
3-fluoroazetidine;hydrochloride | |
10125054 | |
≥98.0% (N,T) |
617718-46-4 | |
White | |
MFCD05663715 | |
3-fluoroazetidine hydrochloride, 3-fluoroazetidine hcl, 3-fluoroazetidinehydrochloride, azetidine, 3-fluoro-, hydrochloride, 3-fluoroazetane hydrochloride, acmc-20dlgf, c3h6fn.hcl, ksc941o8n, 3-fluoro-azetidine hydrochloride, 3-fluoroazetidine hcl salt | |
C1C(CN1)F.Cl | |
111.544 | |
111.54 | |
Crystalline Powder |
Chemical Identifiers
617718-46-4 | |
111.544 | |
PXFUWRWCKSLCLS-UHFFFAOYSA-N | |
10125054 | |
C1C(CN1)F.Cl |
C3H7ClFN | |
MFCD05663715 | |
3-fluoroazetidine hydrochloride, 3-fluoroazetidine hcl, 3-fluoroazetidinehydrochloride, azetidine, 3-fluoro-, hydrochloride, 3-fluoroazetane hydrochloride, acmc-20dlgf, c3h6fn.hcl, ksc941o8n, 3-fluoro-azetidine hydrochloride, 3-fluoroazetidine hcl salt | |
3-fluoroazetidine;hydrochloride |
Safety and Handling
TSCA : No