Learn More
3-Fluorobenzyl isocyanate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 437580010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Fluorobenzyl isocyanate | |
97.5 | |
209.0°C | |
Authentic | |
Glass bottle | |
1g | |
PHRJTGPFEAUEBC-UHFFFAOYSA-N | |
1-fluoro-3-(isocyanatomethyl)benzene | |
643499 | |
98% |
102422-56-0 | |
100.0 | |
91°C | |
97.5% min. (GC) | |
C8H6FNO | |
3-fluorobenzyl isocyanate, 1-fluoro-3-isocyanatomethyl benzene, 3-fluorobenzylisocyanate, benzene,1-fluoro-3-isocyanatomethyl, benzene, 1-fluoro-3-isocyanatomethyl, pubchem16233, acmc-20aov0, 3-fluoro-benzyl isocyanate, 3-fluorophenylmethyl isocyanate | |
C1=CC(=CC(=C1)F)CN=C=O | |
151.14 | |
151.14 |
Chemical Identifiers
102422-56-0 | |
151.14 | |
3-fluorobenzyl isocyanate, 1-fluoro-3-isocyanatomethyl benzene, 3-fluorobenzylisocyanate, benzene,1-fluoro-3-isocyanatomethyl, benzene, 1-fluoro-3-isocyanatomethyl, pubchem16233, acmc-20aov0, 3-fluoro-benzyl isocyanate, 3-fluorophenylmethyl isocyanate | |
1-fluoro-3-(isocyanatomethyl)benzene |
C8H6FNO | |
PHRJTGPFEAUEBC-UHFFFAOYSA-N | |
643499 | |
C1=CC(=CC(=C1)F)CN=C=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reactio
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
RUO â Research Use Only