missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorophenethyl alcohol, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 304690250

 View more versions of this product

Catalog No. AC304690250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Fluorophenethyl alcohol
98.5
1.1200g/mL
107°C
98.5% min. (GC)
C8H9FO
FC6H4CH2CH2OH
25g
2-3-fluorophenyl ethanol, 3-fluorophenethyl alcohol, 2-3-fluorophenyl ethan-1-ol, benzeneethanol, 3-fluoro, 3-fluorophenethylalcohol, 2-3-fluorophenyl ethylalcohol, 3-fluorobenzenethanol, 3-fluorophenylethanol, 3-fluorobenzeneethanol, acmc-209kxl
OCCC1=CC=CC(F)=C1
140.16
140.16
52059-53-7
100.0
90°C (3.0mmHg)
Authentic
Glass bottle
1.5080 to 1.51
MFCD00045998
1.12
MZNBGEKFZCWVES-UHFFFAOYSA-N
2-(3-fluorophenyl)ethanol
573132
99%

Chemical Identifiers

52059-53-7
140.16
MZNBGEKFZCWVES-UHFFFAOYSA-N
573132
OCCC1=CC=CC(F)=C1
C8H9FO
MFCD00045998
2-3-fluorophenyl ethanol, 3-fluorophenethyl alcohol, 2-3-fluorophenyl ethan-1-ol, benzeneethanol, 3-fluoro, 3-fluorophenethylalcohol, 2-3-fluorophenyl ethylalcohol, 3-fluorobenzenethanol, 3-fluorophenylethanol, 3-fluorobenzeneethanol, acmc-209kxl
2-(3-fluorophenyl)ethanol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only