missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoropiperidine hydrochloride, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1977603

 View more versions of this product

Catalog No. AAL1977603


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Fluoropiperidine hydrochloride
∼227°C
MFCD03452799
6145604
3-fluoropiperidine hydrochloride, 3-fluoropiperidine hcl, piperidine, 3-fluoro-, hydrochloride, pubchem24370, acmc-20an39, 3-fluoropiperidinehydrochloride, 3-fluoro-piperidine hydrochloride, 3-fluoranylpiperidine hydrochloride, 3-fluoro-piperidine, hydrochloride
C1CC(CNC1)F.Cl
139.598
139.6
737000-77-0
C5H11ClFN
1 g
Hygroscopic
RDJUBLSLAULIAT-UHFFFAOYSA-N
3-fluoropiperidine;hydrochloride
2759156
97%

Chemical Identifiers

737000-77-0
139.598
RDJUBLSLAULIAT-UHFFFAOYSA-N
2759156
C1CC(CNC1)F.Cl
C5H11ClFN
MFCD03452799
3-fluoropiperidine hydrochloride, 3-fluoropiperidine hcl, piperidine, 3-fluoro-, hydrochloride, pubchem24370, acmc-20an39, 3-fluoropiperidinehydrochloride, 3-fluoro-piperidine hydrochloride, 3-fluoranylpiperidine hydrochloride, 3-fluoro-piperidine, hydrochloride
3-fluoropiperidine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only