missing translation for 'onlineSavingsMsg'
Learn More

3-Furaldehyde, 98%, stabilized

CAS: 498-60-2 | C5H4O2 | 96.08 g/mol

$171.99 - $171.99

Chemical Identifiers

CAS 498-60-2
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.08
InChI Key AZVSIHIBYRHSLB-UHFFFAOYSA-N
Synonym 3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948
PubChem CID 10351
ChEBI CHEBI:87609
IUPAC Name furan-3-carbaldehyde
SMILES C1=COC=C1C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC393260050
SDS View Documents
Thermo Scientific Chemicals
393260050
5 g Glass bottle
Each for $171.99
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

498-60-2
96.08
3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948
CHEBI:87609
C1=COC=C1C=O
C5H4O2
AZVSIHIBYRHSLB-UHFFFAOYSA-N
10351
furan-3-carbaldehyde

Specifications

498-60-2
100.0
1.1500g/mL
48°C
97.5% min. (GC)
C5H4O2
5 g
3-furaldehyde, 3-furancarboxaldehyde, furan-3-carboxaldehyde, 3-formylfuran, 3-furfural, unii-pob632x444, 3furaldehyde, 3-furancarbaldehyde, pubchem6948
C1=COC=C1C=O
96.08
CHEBI:87609
98%
3-Furaldehyde
97.5
Yellow to Brown
145.0°C
Authentic
Glass bottle
1.4930 to 1.4950 (20°C, 589nm)
1.15
AZVSIHIBYRHSLB-UHFFFAOYSA-N
furan-3-carbaldehyde
10351
96.08
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable f

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.- No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathi

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only