missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-Glycidoxypropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

$67.58 - $304.97

Chemical Identifiers

CAS 2530-83-8
Molecular Formula C9H20O5Si
Molecular Weight (g/mol) 236.339
MDL Number MFCD00005144
InChI Key BPSIOYPQMFLKFR-UHFFFAOYSA-N
Synonym 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
PubChem CID 17317
IUPAC Name trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane
SMILES CO[Si](CCCOCC1CO1)(OC)OC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1843114
View Documents
Thermo Scientific Chemicals
A1843114
25 g
Each for $67.58
 
AAA1843122
View Documents
Thermo Scientific Chemicals
A1843122
100 g
Each for $93.51
 
AAA1843136
View Documents
Thermo Scientific Chemicals
A1843136
500 g
Each for $304.97
 
Description

Description

(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.

Solubility
Miscible with alcohols, ketones and aliphatic or aromatic hydrocarbons. Immiscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

2530-83-8
236.339
BPSIOYPQMFLKFR-UHFFFAOYSA-N
17317
CO[Si](CCCOCC1CO1)(OC)OC
C9H20O5Si
MFCD00005144
3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane

Specifications

119°C to 121°C (2mmHg)
2530-83-8
C9H20O5Si
4308125
3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
BPSIOYPQMFLKFR-UHFFFAOYSA-N
trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane
17317
97%
1.429
1.072
<-70°C
25 g
MFCD00005144
135°C (275°F)
Miscible with alcohols,ketones and aliphatic or aromatic hydrocarbons. Immiscible with water.
CO[Si](CCCOCC1CO1)(OC)OC
236.339
236.34
Odorless
Moisture sensitive
(3-Glycidoxypropyl)trimethoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P280-P305+P351+P338-P308+P313-P310-P501c

H318-H341

EINECSNumber : 219-784-2

RTECSNumber : VV4025000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only