missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hepten-2-one, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1908618

 View more versions of this product

Catalog No. AAA1908618


Explore available promotions

Description

Description

It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in most dairy flavors to add greater depth. This ketone will add lingering rich after-notes, to many processed dairy products such as custard, melted butter, creams and cheeses (Brie, Gorgonzola and blue cheeses). Also beneficial in fatty nut applications (brazil and pine nuts, cashews). It is also used to produce Heptan-2-one.

Solubility
Soluble in alcohol and slightly soluble in water(3087 mg/L @ 25°C (est.))

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

3-Hepten-2-one
0.86
49°C (120°F)
1.445
50 g
1739239
Soluble in alcohol and slightly soluble in water(3087mg/L @ 25°C (est.))
CCC\C=C\C(C)=O
112.17
112.16
1119-44-4
156°C
C7H12O
MFCD00015564
UN1224
3-hepten-2-one, butylideneacetone, e-hept-3-en-2-one, 3e-3-hepten-2-one, methyl pentenyl ketone, e-3-hepten-2-one, 3-hepten-2-one, e, unii-tk02t5fjdo, tk02t5fjdo, 3e-hept-3-en-2-one
JHHZQADGLDKIPM-AATRIKPKSA-N
(3E)-hept-3-en-2-one
5364578
97%

Chemical Identifiers

1119-44-4
112.17
JHHZQADGLDKIPM-AATRIKPKSA-N
5364578
C7H12O
MFCD00015564
3-hepten-2-one, butylideneacetone, e-hept-3-en-2-one, 3e-3-hepten-2-one, methyl pentenyl ketone, e-3-hepten-2-one, 3-hepten-2-one, e, unii-tk02t5fjdo, tk02t5fjdo, 3e-hept-3-en-2-one
CCC\C=C\C(C)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 214-278-8

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only