Learn More
3-Heptyne, 97%, Thermo Scientific Chemicals
$248.75 - $248.75
Chemical Identifiers
CAS | 2586-89-2 |
---|---|
Molecular Formula | C7H12 |
Molecular Weight (g/mol) | 96.173 |
MDL Number | MFCD00039960 |
InChI Key | KLYHSJRCIZOUHE-UHFFFAOYSA-N |
Synonym | 3-heptyne, 1-ethyl-2-propylacetylene, acmc-20an2z |
PubChem CID | 75755 |
IUPAC Name | hept-3-yne |
SMILES | CCCC#CCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5350414
|
Thermo Scientific Chemicals
H5350414 |
25 g |
Each for $248.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2586-89-2 | |
96.173 | |
KLYHSJRCIZOUHE-UHFFFAOYSA-N | |
75755 | |
CCCC#CCC |
C7H12 | |
MFCD00039960 | |
3-heptyne, 1-ethyl-2-propylacetylene, acmc-20an2z | |
hept-3-yne |
Specifications
2586-89-2 | |
105°C to 106°C | |
C7H12 | |
CH3(CH2)2C≡CCH2CH3 | |
25 g | |
1731759 | |
KLYHSJRCIZOUHE-UHFFFAOYSA-N | |
hept-3-yne | |
75755 | |
97% |
0.741 | |
−16°C (−3°F) | |
1.4225 | |
MFCD00039960 | |
UN3295 | |
3-heptyne, 1-ethyl-2-propylacetylene, acmc-20an2z | |
CCCC#CCC | |
96.173 | |
96.17 | |
3-Heptyne |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 219-967-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only