missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hexylthiophene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 367380250

 View more versions of this product

Catalog No. AC367380250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Hexylthiophene
97.5
Colorless to Yellow or Red
65.0°C (0.4 mmHg)
Authentic
Glass bottle
1.4945 to 1.4975
25 g
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
CCCCCCC1=CSC=C1
168.3
168.3
Liquid
1693-86-3
100.0
0.9360g/mL
37°C
97.5% min. (GC)
C10H16S
MFCD00143181
0.936
JEDHEMYZURJGRQ-UHFFFAOYSA-N
3-hexylthiophene
566849
98%

Chemical Identifiers

1693-86-3
168.3
JEDHEMYZURJGRQ-UHFFFAOYSA-N
566849
CCCCCCC1=CSC=C1
C10H16S
MFCD00143181
3-n-hexylthiophene, thiophene, 3-hexyl, 3-hexylthiopene, 3-hexylthiophen, 4-hexylthiophene, hexylthiophene 3-, pubchem8861, poly, 3-hexylthiophene, 3-hexylthiophene, 97+%
3-hexylthiophene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Flammable liquid and vapor.
Harmful in contact with skin.

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for brea

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only