missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydrazinobenzoic Acid 97.0+%, TCI America™
Supplier: TCI America H079925G
Specifications
| 3-Hydrazinobenzoic Acid | |
| 183°C | |
| MFCD00191447 | |
| 3-hydrazinobenzoic acid, 3-carboxyphenylhydrazine, 3-hydrazino-benzoic acid, benzoic acid, 3-hydrazino, 3-hydrazinobenzoicacid, 3-hydrazino benzoic acid, acmc-1ctg2 | |
| C1=CC(=CC(=C1)NN)C(=O)O | |
| 152.153 | |
| 152.15 | |
| Crystalline Powder |
| 38235-71-1 | |
| C7H8N2O2 | |
| 25 g | |
| VBYDSMBICNUTKN-UHFFFAOYSA-N | |
| 3-hydrazinylbenzoic acid | |
| 2736521 | |
| ≥97.0% (T) |
Chemical Identifiers
| 38235-71-1 | |
| 152.153 | |
| VBYDSMBICNUTKN-UHFFFAOYSA-N | |
| 2736521 | |
| C1=CC(=CC(=C1)NN)C(=O)O |
| C7H8N2O2 | |
| MFCD00191447 | |
| 3-hydrazinobenzoic acid, 3-carboxyphenylhydrazine, 3-hydrazino-benzoic acid, benzoic acid, 3-hydrazino, 3-hydrazinobenzoicacid, 3-hydrazino benzoic acid, acmc-1ctg2 | |
| 3-hydrazinylbenzoic acid |
Safety and Handling
TSCA : No