missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone 98.0+%, TCI America™
$98.79 - $490.25
Chemical Identifiers
| CAS | 2029-49-4 |
|---|---|
| Molecular Formula | C7H8O5 |
| Molecular Weight (g/mol) | 172.14 |
| MDL Number | MFCD00110732 |
| InChI Key | YZUXYQQFVNGYCA-UHFFFAOYSA-N |
| PubChem CID | 2747691 |
| IUPAC Name | 3-hydroxy-2,6-bis(hydroxymethyl)-4H-pyran-4-one |
| SMILES | OCC1=CC(=O)C(O)=C(CO)O1 |
Chemical Identifiers
| 2029-49-4 | |
| 172.14 | |
| YZUXYQQFVNGYCA-UHFFFAOYSA-N | |
| 3-hydroxy-2,6-bis(hydroxymethyl)-4H-pyran-4-one |
| C7H8O5 | |
| MFCD00110732 | |
| 2747691 | |
| OCC1=CC(=O)C(O)=C(CO)O1 |
Specifications
| 2029-49-4 | |
| C7H8O5 | |
| 1 g | |
| OCC1=CC(=O)C(O)=C(CO)O1 | |
| 172.14 | |
| 172.14 | |
| Crystalline Powder |
| 149°C | |
| MFCD00110732 | |
| YZUXYQQFVNGYCA-UHFFFAOYSA-N | |
| 3-hydroxy-2,6-bis(hydroxymethyl)-4H-pyran-4-one | |
| 2747691 | |
| ≥98.0% (HPLC,T) | |
| 3-Hydroxy-2,6-bis(hydroxymethyl)-4-pyrone |
Safety and Handling
TSCA : No