missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxy-2'-methyl-2-naphthanilide 97.0+%, TCI America™
$132.35 - $1683.24
Chemical Identifiers
| CAS | 135-61-5 |
|---|---|
| Molecular Formula | C18H15NO2 |
| Molecular Weight (g/mol) | 277.323 |
| MDL Number | MFCD00021634 |
| InChI Key | FBLAHUMENIHUGG-UHFFFAOYSA-N |
| Synonym | 2-Hydroxy-3-naphthoic Acid o-Toluidide, Naphthol AS-D, Azoic Coupling Component 18, 3-Hydroxy-N-(o-tolyl)-2-naphthamide |
| PubChem CID | 67273 |
| IUPAC Name | 3-hydroxy-N-(2-methylphenyl)naphthalene-2-carboxamide |
| SMILES | CC1=CC=CC=C1NC(=O)C2=CC3=CC=CC=C3C=C2O |
Chemical Identifiers
| 135-61-5 | |
| 277.323 | |
| FBLAHUMENIHUGG-UHFFFAOYSA-N | |
| 67273 | |
| CC1=CC=CC=C1NC(=O)C2=CC3=CC=CC=C3C=C2O |
| C18H15NO2 | |
| MFCD00021634 | |
| 2-Hydroxy-3-naphthoic Acid o-Toluidide, Naphthol AS-D, Azoic Coupling Component 18, 3-Hydroxy-N-(o-tolyl)-2-naphthamide | |
| 3-hydroxy-N-(2-methylphenyl)naphthalene-2-carboxamide |
Specifications
| 135-61-5 | |
| C18H15NO2 | |
| 25 g | |
| 2-Hydroxy-3-naphthoic Acid o-Toluidide, Naphthol AS-D, Azoic Coupling Component 18, 3-Hydroxy-N-(o-tolyl)-2-naphthamide | |
| CC1=CC=CC=C1NC(=O)C2=CC3=CC=CC=C3C=C2O | |
| 277.323 | |
| 277.32 | |
| Crystalline Powder |
| 198°C | |
| MFCD00021634 | |
| 3077 | |
| FBLAHUMENIHUGG-UHFFFAOYSA-N | |
| 3-hydroxy-N-(2-methylphenyl)naphthalene-2-carboxamide | |
| 67273 | |
| ≥97.0% (HPLC) | |
| 3-Hydroxy-2′-methyl-2-naphthanilide |
Safety and Handling
EINECSNumber : (5)-2286
TSCA : Yes