missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Hydroxy-2-methylbenzoic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1374303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Hydroxy-2-methylbenzoic acid | |
142°C to 146°C | |
C8H7O3 | |
1 g | |
3-hydroxy-o-toluic acid, 3-hydroxy-2-methyl benzoic acid, 3-hydroxy-2-methyl-benzoic acid, 2-methyl-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2-methyl, kresolsaure, 3,2-cresotic acid, pubchem15694, 3-carboxy-2-methylphenol, acmc-1b1jy | |
CC1=C(O)C=CC=C1C([O-])=O | |
151.14 | |
152.15 |
603-80-5 | |
Odorless | |
MFCD00671541 | |
2613791 | |
RIERSGULWXEJKL-UHFFFAOYSA-M | |
3-hydroxy-2-methylbenzoic acid | |
252023 | |
97% |
Chemical Identifiers
603-80-5 | |
151.14 | |
RIERSGULWXEJKL-UHFFFAOYSA-M | |
252023 | |
CC1=C(O)C=CC=C1C([O-])=O |
C8H7O3 | |
MFCD00671541 | |
3-hydroxy-o-toluic acid, 3-hydroxy-2-methyl benzoic acid, 3-hydroxy-2-methyl-benzoic acid, 2-methyl-3-hydroxybenzoic acid, benzoic acid, 3-hydroxy-2-methyl, kresolsaure, 3,2-cresotic acid, pubchem15694, 3-carboxy-2-methylphenol, acmc-1b1jy | |
3-hydroxy-2-methylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only