Learn More
3-Hydroxyazetidine hydrochloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2584206
Description
3-Hydroxyazetidine hydrochloride is an building block used in a recent synthesis of a fluoroquinolone antibiotic and a useful intermediate in the synthesis of polypeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxyazetidine hydrochloride is an building block used in a recent synthesis of a fluoroquinolone antibiotic and a useful intermediate in the synthesis of polypeptides.
Solubility
Soluble in water, DMSO, methanol.
Notes
Hygroscopic. Store under inert gas. Store away from water/ moisture, oxidizing agents.
Specifications
3-Hydroxyazetidine hydrochloride | |
85°C to 90°C | |
MFCD02683887 | |
Hygroscopic | |
Soluble in water,DMSO,methanol. | |
C1C(CN1)O.Cl | |
109.553 | |
109.56 |
18621-18-6 | |
C3H8ClNO | |
5 g | |
3-hydroxyazetidine hydrochloride, azetidin-3-ol hydrochloride, 3-hydroxyazetidine hcl, azetidin-3-ol hcl, 3-azetidinol hydrochloride, azetidin-3-ol hydrochloride salt, 3-azetidinol, hydrochloride, 3-hydroxy-azetidine hydrochloride, azetidin-3-ol, hydrochloride salt, 3-hydroxy-1-azetidine hydrochloride | |
UQUPQEUNHVVNKW-UHFFFAOYSA-N | |
azetidin-3-ol;hydrochloride | |
2759290 | |
97% |
Chemical Identifiers
18621-18-6 | |
109.553 | |
UQUPQEUNHVVNKW-UHFFFAOYSA-N | |
2759290 | |
C1C(CN1)O.Cl |
C3H8ClNO | |
MFCD02683887 | |
3-hydroxyazetidine hydrochloride, azetidin-3-ol hydrochloride, 3-hydroxyazetidine hcl, azetidin-3-ol hcl, 3-azetidinol hydrochloride, azetidin-3-ol hydrochloride salt, 3-azetidinol, hydrochloride, 3-hydroxy-azetidine hydrochloride, azetidin-3-ol, hydrochloride salt, 3-hydroxy-1-azetidine hydrochloride | |
azetidin-3-ol;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only