Learn More
3-Hydroxybenzaldehyde, 97%, Thermo Scientific Chemicals
$44.37 - $451.07
Chemical Identifiers
CAS | 100-83-4 |
---|---|
Molecular Formula | C7H6O2 |
Molecular Weight (g/mol) | 122.123 |
MDL Number | MFCD00003368 |
InChI Key | IAVREABSGIHHMO-UHFFFAOYSA-N |
Synonym | m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde |
PubChem CID | 101 |
ChEBI | CHEBI:16207 |
IUPAC Name | 3-hydroxybenzaldehyde |
SMILES | C1=CC(=CC(=C1)O)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1354114
|
Thermo Scientific Chemicals
A1354114 |
25 g |
Each for $44.37
|
|
||||||
AAA1354122
|
Thermo Scientific Chemicals
A1354122 |
100 g |
Each for $116.14
|
|
||||||
AAA1354136
|
Thermo Scientific Chemicals
A1354136 |
500 g |
Each for $451.07
|
|
||||||
Description
3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Hydroxybenzaldehyde is used as an ionophore, during the development of an highly selective and sensitive PVC membrane sensor, which can be used as a Tb3+ ion selective electrode.
Solubility
Soluble in DMSO and Methanol
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
100-83-4 | |
122.123 | |
IAVREABSGIHHMO-UHFFFAOYSA-N | |
101 | |
3-hydroxybenzaldehyde |
C7H6O2 | |
MFCD00003368 | |
m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde | |
CHEBI:16207 | |
C1=CC(=CC(=C1)O)C=O |
Specifications
100-83-4 | |
191°C (50 mmHg) | |
MFCD00003368 | |
507099 | |
m-formylphenol, m-hydroxybenzaldehyde, 3-formylphenol, benzaldehyde, 3-hydroxy, meta-hydroxybenzaldehyde, benzaldehyde, m-hydroxy, 3-hydroxy-benzaldehyde, 3-hydroxy benzaldehyde, 3-hydroxybenzaldehyd, 3-hydroxylbenzaldehyde | |
IAVREABSGIHHMO-UHFFFAOYSA-N | |
3-hydroxybenzaldehyde | |
101 | |
122.12 | |
3-Hydroxybenzaldehyde |
102°C to 105°C | |
C7H6O2 | |
25 g | |
Air Sensitive | |
Soluble in DMSO and Methanol | |
C1=CC(=CC(=C1)O)C=O | |
122.123 | |
CHEBI:16207 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
EINECSNumber : 202-892-9
RTECSNumber : CU6450000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only