missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Hydroxyphenylacetylene, 97%, Thermo Scientific Chemicals

$171.99 - $616.84

Chemical Identifiers

CAS 10401-11-3
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00078347
InChI Key AODMJIOEGCBUQL-UHFFFAOYSA-N
Synonym 3-hydroxyphenylacetylene, phenol, 3-ethynyl, m-ethynylphenol, 3-ethynyl-phenol, #, 3-hydroxy-phenylacetylene, 3-hydroxyphenyl acetylene, 3-hydroxy-1-ethynylbenzene, phenol, 3-ethynyl-9ci
PubChem CID 139144
IUPAC Name 3-ethynylphenol
SMILES OC1=CC=CC(=C1)C#C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1152303
View Documents
Thermo Scientific Chemicals
L1152303
1 g
Each for $171.99
 
AAL1152306
View Documents
Thermo Scientific Chemicals
L1152306
5 g
Each for $616.84
 
Description

Description

3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyphenylacetylene is used as a fluorogenic and chromogenic probe for bacterial enzymes.

Solubility
Miscible with dimethylformamide.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

10401-11-3
118.14
AODMJIOEGCBUQL-UHFFFAOYSA-N
139144
OC1=CC=CC(=C1)C#C
C8H6O
MFCD00078347
3-hydroxyphenylacetylene, phenol, 3-ethynyl, m-ethynylphenol, 3-ethynyl-phenol, #, 3-hydroxy-phenylacetylene, 3-hydroxyphenyl acetylene, 3-hydroxy-1-ethynylbenzene, phenol, 3-ethynyl-9ci
3-ethynylphenol

Specifications

10401-11-3
231°C
1.587
1 g
3-hydroxyphenylacetylene, phenol, 3-ethynyl, m-ethynylphenol, 3-ethynyl-phenol, #, 3-hydroxy-phenylacetylene, 3-hydroxyphenyl acetylene, 3-hydroxy-1-ethynylbenzene, phenol, 3-ethynyl-9ci
AODMJIOEGCBUQL-UHFFFAOYSA-N
3-ethynylphenol
139144
97%
1.09
C8H6O
MFCD00078347
2076613
Miscible with dimethylformamide.
OC1=CC=CC(=C1)C#C
118.14
118.14
3-Hydroxyphenylacetylene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only