missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodo-1H-pyrazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 444810010

 View more versions of this product

Catalog No. AC444810010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Iodo-1H-pyrazole
96% min. (GC)
C3H3IN2
1g
RUKDVLFJSMVBLV-UHFFFAOYSA-N
5-iodo-1H-pyrazole
1239830
97%
4522-35-4
Glass Bottle
MFCD12022324,MFCD01463487
3-iodo-1h-pyrazole, 3-iodopyrazole, iodopyrazole, 1h-pyrazole, 3-iodo, 3-iodo-2h-pyrazole, iodo-pyrazole, 5-iodopyrazole, acmc-2097sa
IC1=CC=NN1
193.98
193.98

Chemical Identifiers

4522-35-4
193.98
RUKDVLFJSMVBLV-UHFFFAOYSA-N
1239830
IC1=CC=NN1
C3H3IN2
MFCD12022324,MFCD01463487
3-iodo-1h-pyrazole, 3-iodopyrazole, iodopyrazole, 1h-pyrazole, 3-iodo, 3-iodo-2h-pyrazole, iodo-pyrazole, 5-iodopyrazole, acmc-2097sa
5-iodo-1H-pyrazole
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only