missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-nitroanisole, 97%

CAS: 214279-40-0 | C7H6INO3 | 279.03 g/mol

$161.55 - $161.55

Chemical Identifiers

CAS 214279-40-0
Molecular Formula C7H6INO3
Molecular Weight (g/mol) 279.03
MDL Number MFCD01320682
InChI Key NLTAHTYBZZSCIT-UHFFFAOYSA-N
Synonym 3-iodo-4-nitroanisole, 4-nitro-3-iodoanisole, benzene, 2-iodo-4-methoxy-1-nitro, 1-nitro-2-iodo-4-methoxybenzene, 3-iodo-4-nitrophenyl methyl ether, benzene,2-iodo-4-methoxy-1-nitro
PubChem CID 2733456
IUPAC Name 2-iodo-4-methoxy-1-nitrobenzene
SMILES COC1=CC=C(C(I)=C1)[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC326320050
SDS View Documents
Thermo Scientific Chemicals
326320050
5 g Glass bottle
Each for $161.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

214279-40-0
279.03
NLTAHTYBZZSCIT-UHFFFAOYSA-N
2733456
COC1=CC=C(C(I)=C1)[N+]([O-])=O
C7H6INO3
MFCD01320682
3-iodo-4-nitroanisole, 4-nitro-3-iodoanisole, benzene, 2-iodo-4-methoxy-1-nitro, 1-nitro-2-iodo-4-methoxybenzene, 3-iodo-4-nitrophenyl methyl ether, benzene,2-iodo-4-methoxy-1-nitro
2-iodo-4-methoxy-1-nitrobenzene

Specifications

214279-40-0
Authentic
Glass bottle
MFCD01320682
3-iodo-4-nitroanisole, 4-nitro-3-iodoanisole, benzene, 2-iodo-4-methoxy-1-nitro, 1-nitro-2-iodo-4-methoxybenzene, 3-iodo-4-nitrophenyl methyl ether, benzene,2-iodo-4-methoxy-1-nitro
COC1=CC=C(C(I)=C1)[N+]([O-])=O
279.03
279.03
Crystalline Powder or Crystals
68.0°C to 73.0°C
96% min. (GC)
C7H6INO3
5 g
NLTAHTYBZZSCIT-UHFFFAOYSA-N
2-iodo-4-methoxy-1-nitrobenzene
2733456
97%
3-Iodo-4-nitroanisole
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only