missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodo-L-tyrosine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6378906

 View more versions of this product

Catalog No. AAH6378906


Explore available promotions

Description

Description

3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-iodo-l-tyrosine is used in the inhibition of tyrosine hydroxylase. 3-iodo-L-tyrosine is used in the rapid induction of prolactin secretion.

Solubility
Soluble in dilute aqeous acid and dilute aqeous base.

Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat.
Specifications

Specifications

210°C (decomposition)
70-78-0
C9H10INO3
2941266
Soluble in dilute aqeous acid and dilute aqeous base.
N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O
5 g
439744
307.09
98%
3-Iodo-L-tyrosine
White
MFCD00002608
3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
UQTZMGFTRHFAAM-JLDDOWRYNA-N
(2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
307.09
CHEBI:27847
Light sensitive
Powder

Chemical Identifiers

70-78-0
307.09
UQTZMGFTRHFAAM-JLDDOWRYNA-N
439744
(2S)-2-amino-3-(4-hydroxy-3-iodophenyl)propanoic acid
C9H10INO3
MFCD00002608
3-iodo-l-tyrosine, h-tyr 3-i-oh, 3-iodo-tyrosine, monoiodotyrosine, iodotyrosine, 3-monoiodo-l-tyrosine, l-tyrosine, 3-iodo, s-2-amino-3-4-hydroxy-3-iodophenyl propanoic acid, 3-iodotyrosine, 3-iodo-4-hydroxyphenylalanine
CHEBI:27847
N[C@@H](CC1=CC=C(O)C(I)=C1)C(O)=O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-744-8

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only