missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Iodobenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2811506

 View more versions of this product

Catalog No. AAH2811506


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Iodobenzeneboronic acid pinacol ester
70°C to 72°C
MFCD09953498
Light sensitive
HSHFNMSHDHAHDN-UHFFFAOYSA-N
2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
11313483
97%
408492-28-4
C12H16BIO2
5 g
2-3-iodophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodophenylboronic acid pinacol ester, amtb088, 3-iodophenylboronic acid pinacolester, 1,3,2-dioxaborolane,2-3-iodophenyl-4,4,5,5-tetramethyl, 2-3-iodanylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodo-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene
CC1(C)OB(OC1(C)C)C1=CC(I)=CC=C1
329.97
329.97

Chemical Identifiers

408492-28-4
329.97
HSHFNMSHDHAHDN-UHFFFAOYSA-N
11313483
CC1(C)OB(OC1(C)C)C1=CC(I)=CC=C1
C12H16BIO2
MFCD09953498
2-3-iodophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodophenylboronic acid pinacol ester, amtb088, 3-iodophenylboronic acid pinacolester, 1,3,2-dioxaborolane,2-3-iodophenyl-4,4,5,5-tetramethyl, 2-3-iodanylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodo-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene
2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only