Learn More
3-Iodobenzylamine, 97%, Thermo Scientific Chemicals
$188.53 - $522.14
Chemical Identifiers
CAS | 696-40-2 |
---|---|
Molecular Formula | C7H8IN |
Molecular Weight (g/mol) | 233.052 |
MDL Number | MFCD00192227 |
InChI Key | LQLOGZQVKUNBRX-UHFFFAOYSA-N |
Synonym | 3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine |
PubChem CID | 97012 |
IUPAC Name | (3-iodophenyl)methanamine |
SMILES | C1=CC(=CC(=C1)I)CN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1780103
|
Thermo Scientific Chemicals
L1780103 |
1 g |
Each for $188.53
|
|
|||||
AAL1780106
|
Thermo Scientific Chemicals
L1780106 |
5 g |
Each for $522.14
|
|
|||||
Description
It is a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a pharmaceutical intermediate.
Solubility
Difficult to mix in water.
Notes
Air sensitive and light sensitive. Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
Chemical Identifiers
696-40-2 | |
233.052 | |
LQLOGZQVKUNBRX-UHFFFAOYSA-N | |
97012 | |
C1=CC(=CC(=C1)I)CN |
C7H8IN | |
MFCD00192227 | |
3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine | |
(3-iodophenyl)methanamine |
Specifications
696-40-2 | |
132°C (8 mmHg) | |
C7H8IN | |
MFCD00192227 | |
UN2735 | |
Air and light sensitive | |
Difficult to mix in water. | |
C1=CC(=CC(=C1)I)CN | |
233.052 | |
233.05 | |
3-Iodobenzylamine |
1.748 | |
>110°C (230°F) | |
1.6375 | |
1 g | |
2689602 | |
3-iodobenzylamine, 3-iodophenyl methanamine, meta-iodobenzylamine, benzenemethanamine, 3-iodo, 3-iodo-benzylamine, 1-3-iodophenyl methanamine, 3-iodobenzyl amine, m-iodobenzylamine, 3-iodobenzenemethanamine, 3-iodo-benzenemethanamine | |
LQLOGZQVKUNBRX-UHFFFAOYSA-N | |
(3-iodophenyl)methanamine | |
97012 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only