Learn More
3-Iodopropionic acid, 99%, Thermo Scientific Chemicals
$59.58 - $170.55
Chemical Identifiers
CAS | 141-76-4 |
---|---|
Molecular Formula | C3H5IO2 |
Molecular Weight (g/mol) | 199.98 |
MDL Number | MFCD00002765 |
InChI Key | KMRNTNDWADEIIX-UHFFFAOYSA-N |
Synonym | 3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid |
PubChem CID | 8856 |
IUPAC Name | 3-iodopropanoic acid |
SMILES | OC(=O)CCI |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0265506
|
Thermo Scientific Chemicals
L0265506 |
5 g |
Each for $59.58
|
|
|||||
AAL0265514
|
Thermo Scientific Chemicals
L0265514 |
25 g |
Each for $170.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
141-76-4 | |
199.98 | |
KMRNTNDWADEIIX-UHFFFAOYSA-N | |
8856 | |
OC(=O)CCI |
C3H5IO2 | |
MFCD00002765 | |
3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid | |
3-iodopropanoic acid |
Specifications
141-76-4 | |
C3H5IO2 | |
5 g | |
1699514 | |
3-iodopropionic acid, propanoic acid, 3-iodo, beta-iodopropionic acid, propionic acid, 3-iodo, iodopropanoic acid, 3-iodo-propionic acid, .beta.-iodopropionic acid, iodo-propanoic acid, b-iodopropionic acid, 3-iodo-propanoic acid | |
OC(=O)CCI | |
199.98 | |
199.98 | |
3-Iodopropionic acid |
80°C to 83°C | |
MFCD00002765 | |
UN3261 | |
Light sensitive | |
KMRNTNDWADEIIX-UHFFFAOYSA-N | |
3-iodopropanoic acid | |
8856 | |
99% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 205-499-0
RTECSNumber : UF4900000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only