missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Mercapto-3-methyl-1-butanol 98.0+%, TCI America™
$223.97 - $711.71
Chemical Identifiers
CAS | 34300-94-2 |
---|---|
Molecular Formula | C5H12OS |
Molecular Weight (g/mol) | 120.21 |
MDL Number | MFCD00101881 |
InChI Key | GBCGIJAYTBMFHI-UHFFFAOYSA-N |
PubChem CID | 520682 |
IUPAC Name | 3-methyl-3-sulfanylbutan-1-ol |
SMILES | CC(C)(S)CCO |
Chemical Identifiers
34300-94-2 | |
120.21 | |
GBCGIJAYTBMFHI-UHFFFAOYSA-N | |
3-methyl-3-sulfanylbutan-1-ol |
C5H12OS | |
MFCD00101881 | |
520682 | |
CC(C)(S)CCO |
Specifications
34300-94-2 | |
90°C | |
MFCD00101881 | |
GBCGIJAYTBMFHI-UHFFFAOYSA-N | |
3-methyl-3-sulfanylbutan-1-ol | |
520682 | |
≥98.0% (GC,T) | |
3-Mercapto-3-methyl-1-butanol |
Colorless | |
C5H12OS | |
5 g | |
CC(C)(S)CCO | |
120.21 | |
120.21 | |
Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator