Learn More
(3-Mercaptopropyl)trimethoxysilane, 85%, Tech.
CAS: 4420-74-0 | C6H16O3SSi | 196.34 g/mol
$95.61 - $315.93
Chemical Identifiers
| CAS | 4420-74-0 |
|---|---|
| Molecular Formula | C6H16O3SSi |
| Molecular Weight (g/mol) | 196.34 |
| MDL Number | MFCD00004901 |
| InChI Key | UUEWCQRISZBELL-UHFFFAOYSA-N |
| Synonym | trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810 |
| PubChem CID | 20473 |
| IUPAC Name | 3-trimethoxysilylpropane-1-thiol |
| SMILES | CO[Si](CCCS)(OC)OC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC174650500
|
Thermo Scientific Chemicals
174650500 |
50 g | Glass bottle |
Each for $95.61
|
|
||||
|
AC174652500
|
Thermo Scientific Chemicals
174652500 |
250 g | Glass bottle |
Each for $315.93
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4420-74-0 | |
| 196.34 | |
| UUEWCQRISZBELL-UHFFFAOYSA-N | |
| 20473 | |
| CO[Si](CCCS)(OC)OC |
| C6H16O3SSi | |
| MFCD00004901 | |
| trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810 | |
| 3-trimethoxysilylpropane-1-thiol |
Specifications
| 4420-74-0 | |
| 100.0 | |
| 1.0500g/mL | |
| 88°C | |
| 80% min. (GC) | |
| C6H16O3SSi | |
| HSCH2CH2CH2Si(OCH3)3 | |
| 50 g | |
| 1.05 | |
| Solubility in water: decomposes. Other solubilities: soluble in methanol,ethanol,isopropanol,,mineral spirits,acetone,benzene,toluene and,xylene | |
| CO[Si](CCCS)(OC)OC | |
| 196.34 | |
| 2 mPa.s ( 20°C) | |
| 85% | |
| Liquid |
| 80.0 | |
| Colorless to Yellow | |
| 215°C | |
| Authentic | |
| Glass bottle | |
| 1.4410 to 1.443 | |
| MFCD00004901 | |
| 10, 148A | |
| trimethoxysilylpropanethiol, 3-mercaptopropyltrimethoxysilane, 3-mercaptopropyl trimethoxysilane, 1-propanethiol, 3-trimethoxysilyl, silquest a 189, 3-trimethoxysilyl propanethiol, prosil 196, silane a 189, union carbide a-189, sila-ace s 810 | |
| UUEWCQRISZBELL-UHFFFAOYSA-N | |
| 3-trimethoxysilylpropane-1-thiol | |
| 20473 | |
| 196.34 | |
| Technical | |
| (3-Mercaptopropyl)trimethoxysilane |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 224-588-5
RUO – Research Use Only