Learn More
3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane, 98%, Thermo Scientific Chemicals
$113.77 - $392.63
Chemical Identifiers
CAS | 17096-07-0 |
---|---|
Molecular Formula | C16H38O5Si4 |
Molecular Weight (g/mol) | 422.815 |
MDL Number | MFCD00053871 |
InChI Key | BESKSSIEODQWBP-UHFFFAOYSA-N |
Synonym | 3-methacryloyloxy propyltris trimethylsiloxy silane, 3-tris trimethylsiloxy silyl propyl methacrylate, 3-1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxan-3-yl propyl methacrylate, unii-d32s78j7t8, 3-methacryloyloxypropyl tris trimethylsiloxy silane, 3-methacryloxypropyltris trimethylsiloxy silane, 3-tris trimethylsilyloxy silyl propyl methacrylate, methacryloxypropyltris trimethylsiloxy silane, 3-3,3,3-trimethyl-1,1-bis trimethylsilyl oxy disiloxanyl propyl methacrylate |
PubChem CID | 123371 |
IUPAC Name | 3-tris(trimethylsilyloxy)silylpropyl 2-methylprop-2-enoate |
SMILES | CC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL1668214
|
Thermo Scientific Chemicals
L1668214 |
25 g |
Each for $113.77
|
|
||||||
AAL1668222
|
Thermo Scientific Chemicals
L1668222 |
100 g |
Each for $392.63
|
|
||||||
Description
3-(Methacryloyloxy)propyltris(trimethylsiloxy) silane acts as a tetramethylsilane-protected silicate compound utilized for proteomics research. It is also used as a silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent and silane crosslinking agent. It is also employed as a silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent and silane reinforcer. Further, it serves as a silylating agent, silane reducing agent, silyl building block and synthons in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(Methacryloyloxy)propyltris(trimethylsiloxy) silane acts as a tetramethylsilane-protected silicate compound utilized for proteomics research. It is also used as a silane coupling agent, silane adhesion promoter, silane hydrophobing agent, silane dispersing agent and silane crosslinking agent. It is also employed as a silane moisture scavenger, polypropylene catalyst, silicate stabilizer, polyurethane endcapper silane, silane drying agent, silane curing agent and silane reinforcer. Further, it serves as a silylating agent, silane reducing agent, silyl building block and synthons in synthetic chemistry.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agent.
Chemical Identifiers
17096-07-0 | |
422.815 | |
BESKSSIEODQWBP-UHFFFAOYSA-N | |
123371 | |
CC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C |
C16H38O5Si4 | |
MFCD00053871 | |
3-methacryloyloxy propyltris trimethylsiloxy silane, 3-tris trimethylsiloxy silyl propyl methacrylate, 3-1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxan-3-yl propyl methacrylate, unii-d32s78j7t8, 3-methacryloyloxypropyl tris trimethylsiloxy silane, 3-methacryloxypropyltris trimethylsiloxy silane, 3-tris trimethylsilyloxy silyl propyl methacrylate, methacryloxypropyltris trimethylsiloxy silane, 3-3,3,3-trimethyl-1,1-bis trimethylsilyl oxy disiloxanyl propyl methacrylate | |
3-tris(trimethylsilyloxy)silylpropyl 2-methylprop-2-enoate |
Specifications
112°C to 115°C (0.2mmHg) | |
25 g | |
MFCD00053871 | |
3-methacryloyloxy propyltris trimethylsiloxy silane, 3-tris trimethylsiloxy silyl propyl methacrylate, 3-1,1,1,5,5,5-hexamethyl-3-trimethylsilyl oxy trisiloxan-3-yl propyl methacrylate, unii-d32s78j7t8, 3-methacryloyloxypropyl tris trimethylsiloxy silane, 3-methacryloxypropyltris trimethylsiloxy silane, 3-tris trimethylsilyloxy silyl propyl methacrylate, methacryloxypropyltris trimethylsiloxy silane, 3-3,3,3-trimethyl-1,1-bis trimethylsilyl oxy disiloxanyl propyl methacrylate | |
BESKSSIEODQWBP-UHFFFAOYSA-N | |
3-tris(trimethylsilyloxy)silylpropyl 2-methylprop-2-enoate | |
123371 | |
98% | |
0.93 |
17096-07-0 | |
C16H38O5Si4 | |
201°C (393°F) | |
Immiscible with water. | |
CC(=C)C(=O)OCCC[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C | |
422.815 | |
422.82 | |
1.4175 | |
3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 241-165-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only