Learn More
3-Methoxy-2-methylbenzoic acid, 97%
CAS: 55289-06-0 | C9H10O3 | 166.18 g/mol
$47.14 - $157.27
Chemical Identifiers
| CAS | 55289-06-0 |
|---|---|
| Molecular Formula | C9H10O3 |
| Molecular Weight (g/mol) | 166.18 |
| MDL Number | MFCD02094039 |
| InChI Key | JPCISVSOTKMFPG-UHFFFAOYSA-N |
| Synonym | 3-methoxy-o-toluic acid, 2-methyl-m-anisic acid, 3-methoxy-2-methyl-benzoic acid, 3-methoxy-2-methylbenzoicacid, benzoic acid, 3-methoxy-2-methyl, 2-methyl-3-methoxybenzoic acid, pubchem4976, acmc-209lm1, ksc269c9h, 2-methyl-3-methoxy-benzoic acid |
| PubChem CID | 7021483 |
| IUPAC Name | 3-methoxy-2-methylbenzoic acid |
| SMILES | COC1=CC=CC(C(O)=O)=C1C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1872909
|
Thermo Scientific Chemicals
L1872909 |
10 g |
Each for $47.14
|
|
|||||
|
AAL1872918
|
Thermo Scientific Chemicals
L1872918 |
50 g |
Each for $157.27
|
|
|||||
Description
3-Methoxy-2-methylbenzoic acid is used as a Corey-Bakshi-Shibata oxazaborolidine catalyst for asymmetric reduction and asymmetric synthesis. It is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols and propargyl alcohols. It is also used in desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methoxy-2-methylbenzoic acid is used as a Corey-Bakshi-Shibata oxazaborolidine catalyst for asymmetric reduction and asymmetric synthesis. It is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols and propargyl alcohols. It is also used in desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
| 55289-06-0 | |
| 166.18 | |
| JPCISVSOTKMFPG-UHFFFAOYSA-N | |
| 7021483 | |
| COC1=CC=CC(C(O)=O)=C1C |
| C9H10O3 | |
| MFCD02094039 | |
| 3-methoxy-o-toluic acid, 2-methyl-m-anisic acid, 3-methoxy-2-methyl-benzoic acid, 3-methoxy-2-methylbenzoicacid, benzoic acid, 3-methoxy-2-methyl, 2-methyl-3-methoxybenzoic acid, pubchem4976, acmc-209lm1, ksc269c9h, 2-methyl-3-methoxy-benzoic acid | |
| 3-methoxy-2-methylbenzoic acid |
Specifications
| 55289-06-0 | |
| MFCD02094039 | |
| 510395 | |
| Soluble in methanol. Insoluble in water. | |
| COC1=CC=CC(C(O)=O)=C1C | |
| 166.18 | |
| 166.18 | |
| 3-Methoxy-2-methylbenzoic acid |
| C9H10O3 | |
| 10 g | |
| 3-methoxy-o-toluic acid, 2-methyl-m-anisic acid, 3-methoxy-2-methyl-benzoic acid, 3-methoxy-2-methylbenzoicacid, benzoic acid, 3-methoxy-2-methyl, 2-methyl-3-methoxybenzoic acid, pubchem4976, acmc-209lm1, ksc269c9h, 2-methyl-3-methoxy-benzoic acid | |
| JPCISVSOTKMFPG-UHFFFAOYSA-N | |
| 3-methoxy-2-methylbenzoic acid | |
| 7021483 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only