Learn More
3-Methoxy-3-methyl-1-butanol, 98+%, Thermo Scientific Chemicals
$50.93 - $478.92
Chemical Identifiers
CAS | 56539-66-3 |
---|---|
Molecular Formula | C6H14O2 |
Molecular Weight (g/mol) | 118.176 |
MDL Number | MFCD00044771 |
InChI Key | MFKRHJVUCZRDTF-UHFFFAOYSA-N |
Synonym | 3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389 |
PubChem CID | 62118 |
IUPAC Name | 3-methoxy-3-methylbutan-1-ol |
SMILES | CC(C)(CCO)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL15953AE
|
Thermo Scientific Chemicals
L15953AE |
100 mL |
Each for $50.93
|
|
|||||
AAL15953AP
|
Thermo Scientific Chemicals
L15953AP |
500 mL |
Each for $134.33
|
|
|||||
AAL159530F
|
Thermo Scientific Chemicals
L159530F |
2500 mL |
Each for $478.92
|
|
|||||
Description
3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methoxy-3-methyl-1-butanol is a solvent that compares favorably with its isomer ethylene glycol monobutyl ether. It is employed as raw material for the production of industrial detergents. It is also used as solvent for paints, inks, and fragrances.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Chemical Identifiers
56539-66-3 | |
118.176 | |
MFKRHJVUCZRDTF-UHFFFAOYSA-N | |
62118 | |
CC(C)(CCO)OC |
C6H14O2 | |
MFCD00044771 | |
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389 | |
3-methoxy-3-methylbutan-1-ol |
Specifications
56539-66-3 | |
173°C to 175°C | |
C6H14O2 | |
MFCD00044771 | |
1732882 | |
Soluble in water. | |
CC(C)(CCO)OC | |
118.176 | |
118.18 | |
3-Methoxy-3-methyl-1-butanol |
0.926 | |
71°C (159°F) | |
1.428 | |
100 mL | |
3-methoxy-3-methyl-1-butanol, 3-methyl-3-methoxybutanol, 3-methoxy-3-methylbutanol, 1-butanol, 3-methoxy-3-methyl, methoxymethylbutanol, 3-methoxy-3-methyl-butan-1-ol, acmc-1bbqk, dsstox_cid_24389, dsstox_rid_80190, dsstox_gsid_44389 | |
MFKRHJVUCZRDTF-UHFFFAOYSA-N | |
3-methoxy-3-methylbutan-1-ol | |
62118 | |
≥98% |
Safety and Handling
GHS H Statement
H227
Combustible liquid.
P210-P264b-P280-P305+P351+P338-P370+P378q-P501c
H227-H319
EINECSNumber : 260-252-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only