Learn More
3-Methoxy-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
$97.26 - $344.23
Chemical Identifiers
CAS | 41789-95-1 |
---|---|
Molecular Formula | C9H14NO |
Molecular Weight (g/mol) | 152.22 |
MDL Number | MFCD04618244 |
InChI Key | FIFKRPFWLHBMHL-UHFFFAOYSA-O |
Synonym | 3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine |
PubChem CID | 5152225 |
IUPAC Name | 1-(3-methoxyphenyl)-N-methylmethanamine |
SMILES | C[NH2+]CC1=CC=CC(OC)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5497003
|
Thermo Scientific Chemicals
H5497003 |
1 g |
Each for $97.26
|
|
|||||
AAH5497006
|
Thermo Scientific Chemicals
H5497006 |
5 g |
Each for $344.23
|
|
|||||
Description
3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.
Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Chemical Identifiers
41789-95-1 | |
152.22 | |
FIFKRPFWLHBMHL-UHFFFAOYSA-O | |
5152225 | |
C[NH2+]CC1=CC=CC(OC)=C1 |
C9H14NO | |
MFCD04618244 | |
3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine | |
1-(3-methoxyphenyl)-N-methylmethanamine |
Specifications
41789-95-1 | |
130°C to 133°C | |
C9H14NO | |
MFCD04618244 | |
Air Sensitive | |
Soluble in chloroform,dichloromethane,ethyl acetate and methanol. | |
C[NH2+]CC1=CC=CC(OC)=C1 | |
152.22 | |
151.21 | |
3-Methoxy-N-methylbenzylamine |
1.014 | |
100°C (212°F) | |
1.529 | |
1 g | |
3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine | |
FIFKRPFWLHBMHL-UHFFFAOYSA-O | |
1-(3-methoxyphenyl)-N-methylmethanamine | |
5152225 | |
97% |
Safety and Handling
GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P264b-P270-P280-P285-P301+P312-P304+P341-P305+P351+P338-P310-P330-P342+P311-P501c
H302-H318-H334
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only