Learn More
3-Methoxy-N,N-dimethylaniline, 98%, Thermo Scientific Chemicals
$156.52 - $156.52
Chemical Identifiers
CAS | 15799-79-8 |
---|---|
Molecular Formula | C9H13NO |
Molecular Weight (g/mol) | 151.209 |
MDL Number | MFCD00051779 |
InChI Key | MOYHVSKDHLMMPS-UHFFFAOYSA-N |
Synonym | 3-dimethylaminoanisole, n,n-dimethyl-m-anisidine, 3-methoxy-n,n-dimethylbenzenamine, 3-methoxyphenyl dimethylamine, 3-dimethylamino-anisole, acmc-1c3gj, m-methoxy-n,n-dimethylanilin, n,n-dimethyl-3-methoxyaniline, n1,n1-dimethyl-3-methoxyaniline, 3-methoxy-n,n-dimethylbenzeneamine |
PubChem CID | 139977 |
IUPAC Name | 3-methoxy-N,N-dimethylaniline |
SMILES | CN(C)C1=CC(=CC=C1)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0870009
|
Thermo Scientific Chemicals
L0870009 |
10 g |
Each for $156.52
|
|
|||||
Description
N,N-Dimethyl-m-anisidine is a phenyl amide with antineoplastic and antibacterial properties. It can be used to produce C20H19NO4. It will need reagent tributyl orthoformate and solvent propan-2-ol with reaction time of 90 min.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN,N-Dimethyl-m-anisidine is a phenyl amide with antineoplastic and antibacterial properties. It can be used to produce C20H19NO4. It will need reagent tributyl orthoformate and solvent propan-2-ol with reaction time of 90 min.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
15799-79-8 | |
151.209 | |
MOYHVSKDHLMMPS-UHFFFAOYSA-N | |
139977 | |
CN(C)C1=CC(=CC=C1)OC |
C9H13NO | |
MFCD00051779 | |
3-dimethylaminoanisole, n,n-dimethyl-m-anisidine, 3-methoxy-n,n-dimethylbenzenamine, 3-methoxyphenyl dimethylamine, 3-dimethylamino-anisole, acmc-1c3gj, m-methoxy-n,n-dimethylanilin, n,n-dimethyl-3-methoxyaniline, n1,n1-dimethyl-3-methoxyaniline, 3-methoxy-n,n-dimethylbenzeneamine | |
3-methoxy-N,N-dimethylaniline |
Specifications
15799-79-8 | |
113°C to 114°C (6 mmHg) | |
1.559 | |
10 g | |
1423591 | |
Not miscible or difficult to mix in water. | |
CN(C)C1=CC(=CC=C1)OC | |
151.209 | |
151.21 | |
3-Methoxy-N,N-dimethylaniline |
1.031 | |
C9H13NO | |
MFCD00051779 | |
UN2810 | |
3-dimethylaminoanisole, n,n-dimethyl-m-anisidine, 3-methoxy-n,n-dimethylbenzenamine, 3-methoxyphenyl dimethylamine, 3-dimethylamino-anisole, acmc-1c3gj, m-methoxy-n,n-dimethylanilin, n,n-dimethyl-3-methoxyaniline, n1,n1-dimethyl-3-methoxyaniline, 3-methoxy-n,n-dimethylbenzeneamine | |
MOYHVSKDHLMMPS-UHFFFAOYSA-N | |
3-methoxy-N,N-dimethylaniline | |
139977 | |
98% |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only