Learn More
3-Methoxyazetidine hydrochloride, 95%
CAS: 148644-09-1 | C4H10ClNO | 123.58 g/mol
Supplier: Thermo Scientific Chemicals H57769ME
Description
It is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the preparation of isoxazole-thiazole derivatives as GABA. It is a receptor inverse agonists for use in the treatment of cognitive disorders.
Notes
Air sensitive. Store in a cool ventilated place. Avoid strong oxidizing materials and heat.
Spécifications
| 3-Methoxyazetidine hydrochloride | |
| 143°C to 149°C | |
| MFCD06804514 | |
| 3-methoxyazetidine hydrochloride, 3-methoxy-azetidine hydrochloride, 3-azetidinyl methyl ether hydrochloride, 3-methoxyazetidine hcl, azetidine, 3-methoxy-, hydrochloride, azetidine, 3-methoxy-,hydrochloride 1:1, acmc-20a0d9, ksc495e1t, 3-methoxyazetidinehydrochloride, 3-methoxy-azetidine-hydrochloride | |
| COC1CNC1.Cl | |
| 123.58 | |
| 123.58 |
| 148644-09-1 | |
| C4H10ClNO | |
| 500 mg | |
| KSXGQRBTBLQJEF-UHFFFAOYSA-N | |
| 3-methoxyazetidine;hydrochloride | |
| 22242858 | |
| 95% |
Identifiants chimiques
| 148644-09-1 | |
| 123.58 | |
| KSXGQRBTBLQJEF-UHFFFAOYSA-N | |
| 22242858 | |
| COC1CNC1.Cl |
| C4H10ClNO | |
| MFCD06804514 | |
| 3-methoxyazetidine hydrochloride, 3-methoxy-azetidine hydrochloride, 3-azetidinyl methyl ether hydrochloride, 3-methoxyazetidine hcl, azetidine, 3-methoxy-, hydrochloride, azetidine, 3-methoxy-,hydrochloride 1:1, acmc-20a0d9, ksc495e1t, 3-methoxyazetidinehydrochloride, 3-methoxy-azetidine-hydrochloride | |
| 3-methoxyazetidine;hydrochloride |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only