missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3'-Methoxybiphenyl-4-sulfonyl chloride, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3321106

 View more versions of this product

Catalog No. AAH3321106


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3'-Methoxybiphenyl-4-sulfonyl chloride
80°C to 84°C
MFCD06409270
UN3261
3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
282.738
282.74
186550-26-5
C13H11ClO3S
5 g
Moisture sensitive
DVQBUJTYQXKLKZ-UHFFFAOYSA-N
4-(3-methoxyphenyl)benzenesulfonyl chloride
10826798
96%

Chemical Identifiers

186550-26-5
282.738
DVQBUJTYQXKLKZ-UHFFFAOYSA-N
10826798
COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl
C13H11ClO3S
MFCD06409270
3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
4-(3-methoxyphenyl)benzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only