missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methoxybutyl 3-Mercaptopropionate 97.0+%, TCI America™
$26.54 - $235.57
Chemical Identifiers
CAS | 27431-40-9 |
---|---|
Molecular Formula | C8H16O3S |
Molecular Weight (g/mol) | 192.273 |
MDL Number | MFCD04038137 |
InChI Key | APWBGRBFKMJPLW-UHFFFAOYSA-N |
Synonym | 3-Mercaptopropionic Acid 3-Methoxybutyl Ester |
PubChem CID | 122398 |
IUPAC Name | 3-methoxybutyl 3-sulfanylpropanoate |
SMILES | CC(CCOC(=O)CCS)OC |
Chemical Identifiers
27431-40-9 | |
192.273 | |
APWBGRBFKMJPLW-UHFFFAOYSA-N | |
122398 | |
CC(CCOC(=O)CCS)OC |
C8H16O3S | |
MFCD04038137 | |
3-Mercaptopropionic Acid 3-Methoxybutyl Ester | |
3-methoxybutyl 3-sulfanylpropanoate |
Specifications
27431-40-9 | |
C8H16O3S | |
25 g | |
APWBGRBFKMJPLW-UHFFFAOYSA-N | |
3-methoxybutyl 3-sulfanylpropanoate | |
122398 | |
≥97.0% (GC,T) | |
3-Methoxybutyl 3-Mercaptopropionate |
Yellow | |
MFCD04038137 | |
3-Mercaptopropionic Acid 3-Methoxybutyl Ester | |
CC(CCOC(=O)CCS)OC | |
192.273 | |
192.27 | |
Liquid |
Safety and Handling
TSCA : No