missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2744403

 View more versions of this product

Catalog No. AAH2744403


Explore available promotions

Description

Description

Reagent used for tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, one-pot ipso-nitration of arylboronic acids, copper-catalyzed nitration, cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling. Reagent used in Preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid6, chromenones and their bradykinin B1 antagonistic active, Pt nanoparticles at Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injecti, salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used for tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, one-pot ipso-nitration of arylboronic acids, copper-catalyzed nitration, cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling. Reagent used in Preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid6, chromenones and their bradykinin B1 antagonistic active, Pt nanoparticles at Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injecti, salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

3-(Methoxycarbonyl)benzeneboronic acid
136°C
MFCD02093046
3-methoxycarbonyl phenylboronic acid, 3-methoxycarbonylphenyl boronic acid, 3-methoxycarbonyl phenyl boronic acid, 3-methoxycarbonyl benzeneboronic acid, 3-methoxycarbonylphenylbaronic acid, methyl 3-dihydroxyboranyl benzoate, m-methoxycarbonyl phenylboronic acid, 3-carbomethoxy-phenylboronic acid
ALTLCJHSJMGSLT-UHFFFAOYSA-N
[3-(methoxycarbonyl)phenyl]boronic acid
2734714
97%
99769-19-4
C8H9BO4
1 g
Reacts with water.
COC(=O)C1=CC=CC(=C1)B(O)O
179.97
179.97

Chemical Identifiers

99769-19-4
179.97
ALTLCJHSJMGSLT-UHFFFAOYSA-N
2734714
C8H9BO4
MFCD02093046
3-methoxycarbonyl phenylboronic acid, 3-methoxycarbonylphenyl boronic acid, 3-methoxycarbonyl phenyl boronic acid, 3-methoxycarbonyl benzeneboronic acid, 3-methoxycarbonylphenylbaronic acid, methyl 3-dihydroxyboranyl benzoate, m-methoxycarbonyl phenylboronic acid, 3-carbomethoxy-phenylboronic acid
COC(=O)C1=CC=CC(=C1)B(O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only