Learn More
3-Methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1095114
Description
A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.
Solubility
Soluble in Chloroform and Ethyl Acetate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
3-Methoxyphenylacetic acid | |
67°C to 72°C | |
MFCD00004334 | |
2614004 | |
Soluble in Chloroform and Ethyl Acetate. | |
COC1=CC=CC(=C1)CC(=O)O | |
166.176 | |
166.18 |
1798-09-0 | |
C9H10O3 | |
25 g | |
3-methoxyphenylacetic acid, 2-3-methoxyphenyl acetic acid, m-methoxyphenylacetic acid, 3-methoxyphenyl acetic acid, 3-methoxybenzeneacetic acid, benzeneacetic acid, 3-methoxy, acetic acid, m-methoxyphenyl, unii-25xlo0t6my, 3-methoxy-phenyl-acetic acid, 25xlo0t6my | |
LEGPZHPSIPPYIO-UHFFFAOYSA-N | |
2-(3-methoxyphenyl)acetic acid | |
15719 | |
97% |
Chemical Identifiers
1798-09-0 | |
166.176 | |
LEGPZHPSIPPYIO-UHFFFAOYSA-N | |
15719 | |
COC1=CC=CC(=C1)CC(=O)O |
C9H10O3 | |
MFCD00004334 | |
3-methoxyphenylacetic acid, 2-3-methoxyphenyl acetic acid, m-methoxyphenylacetic acid, 3-methoxyphenyl acetic acid, 3-methoxybenzeneacetic acid, benzeneacetic acid, 3-methoxy, acetic acid, m-methoxyphenyl, unii-25xlo0t6my, 3-methoxy-phenyl-acetic acid, 25xlo0t6my | |
2-(3-methoxyphenyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-282-8
RTECSNumber : AI8940000
TSCA : No
RUO – Research Use Only