missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxyphenylacetone, 97%

CAS: 3027-13-2 | C10H12O2 | 164.20 g/mol

$153.22 - $598.44

Chemical Identifiers

CAS 3027-13-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008771
InChI Key RMMRRRLPDBJBQL-UHFFFAOYSA-N
Synonym 3-methoxyphenylacetone, 1-3-methoxyphenyl propan-2-one, 1-3-methoxyphenyl acetone, 3-methoxyphenyl acetone, m-methoxyphenyl acetone, m-methoxyphenylacetone, 2-propanone, 1-3-methoxyphenyl, 3-methoxyphenyl methyl ketone, acmc-1ai0k
PubChem CID 76410
IUPAC Name 1-(3-methoxyphenyl)propan-2-one
SMILES COC1=CC=CC(CC(C)=O)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2091603
View Documents
Thermo Scientific Chemicals
B2091603
1 g
Each for $153.22
Only null left
 
AAB2091606
View Documents
Thermo Scientific Chemicals
B2091606
5 g
Each for $598.44
Only null left
 
Description

Description

3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxyphenylacetone is employed in the preparation of optically active cyanohydrins. It is also used in the synthesis of central nervous system (CNS) active compounds. Further, it plays an important role in the preparation of 4-amino-3-methoxypropiophenone.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents, strong acids and bases.
Specifications

Chemical Identifiers

3027-13-2
164.20
RMMRRRLPDBJBQL-UHFFFAOYSA-N
76410
COC1=CC=CC(CC(C)=O)=C1
C10H12O2
MFCD00008771
3-methoxyphenylacetone, 1-3-methoxyphenyl propan-2-one, 1-3-methoxyphenyl acetone, 3-methoxyphenyl acetone, m-methoxyphenyl acetone, m-methoxyphenylacetone, 2-propanone, 1-3-methoxyphenyl, 3-methoxyphenyl methyl ketone, acmc-1ai0k
1-(3-methoxyphenyl)propan-2-one

Specifications

3027-13-2
258°C to 260°C
C10H12O2
MFCD00008771
2044357
Miscible with dimethyl sulfoxide.
COC1=CC=CC(CC(C)=O)=C1
164.20
164.2
3-Methoxyphenylacetone
1.078
>110°C (230°F)
1.5252
1 g
3-methoxyphenylacetone, 1-3-methoxyphenyl propan-2-one, 1-3-methoxyphenyl acetone, 3-methoxyphenyl acetone, m-methoxyphenyl acetone, m-methoxyphenylacetone, 2-propanone, 1-3-methoxyphenyl, 3-methoxyphenyl methyl ketone, acmc-1ai0k
RMMRRRLPDBJBQL-UHFFFAOYSA-N
1-(3-methoxyphenyl)propan-2-one
76410
97%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-191-9

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only