missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals

$164.76 - $164.76

Chemical Identifiers

CAS 36282-40-3
Molecular Formula C7H7BrMgO
Molecular Weight (g/mol) 211.34
MDL Number MFCD00672002
InChI Key AJKNYEHIJAMGHX-UHFFFAOYSA-M
Synonym 3-methoxyphenylmagnesium bromide, grignard reagent, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromid, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, m-methoxyphenyl magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide
PubChem CID 4605969
SMILES COC1=CC=CC([Mg]Br)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC426751000
SDS View Documents
Thermo Scientific Chemicals
426751000
100 mL AcroSeal™ Glass bottle
Each for $164.76
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

36282-40-3
211.34
AJKNYEHIJAMGHX-UHFFFAOYSA-M
4605969
C7H7BrMgO
MFCD00672002
3-methoxyphenylmagnesium bromide, grignard reagent, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromid, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, m-methoxyphenyl magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide
COC1=CC=CC([Mg]Br)=C1

Specifications

36282-40-3,109-99-9,108-88-3
23.25,80.98
1.0000g/mL
AcroSeal™ Glass bottle
CH3OC6H4MgBr
100 mL
1
AJKNYEHIJAMGHX-UHFFFAOYSA-M
magnesium;methoxybenzene;bromide
4605969
Solution
19.02,76.75
Yellow to Brown
−21°C
C7H7BrMgO
MFCD00672002
2924
3-methoxyphenylmagnesium bromide, grignard reagent, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromid, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, m-methoxyphenyl magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide
COC1=CC=CC([Mg]Br)=C1
211.34
211.34
3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
May be fatal if swallowed and enters airways.
May c

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only