Learn More
3-Methyl-1-butanol, 99%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals
$153.69 - $153.69
Chemical Identifiers
CAS | 123-51-3 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
InChI Key | PHTQWCKDNZKARW-UHFFFAOYSA-N |
Synonym | 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol |
PubChem CID | 31260 |
ChEBI | CHEBI:15837 |
IUPAC Name | 3-methylbutan-1-ol |
SMILES | CC(C)CCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC412720010
|
Thermo Scientific Chemicals
412720010 |
1 L | AcroSeal™ Glass bottle |
Each for $153.69
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-51-3 | |
88.15 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CHEBI:15837 | |
CC(C)CCO |
C5H12O | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
31260 | |
3-methylbutan-1-ol |
Specifications
123-51-3 | |
0.8000g/mL | |
45°C | |
AcroSeal™ Glass bottle | |
1.4050 to 1.4070 | |
1 L | |
0.8 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
3-methylbutan-1-ol | |
31260 | |
CHEBI:15837 | |
99% | |
3-Methyl-1-butanol |
-70.0°C | |
130.0°C | |
98.5% min. (GC) | |
C5H12O | |
(CH3)2CHCH2CH2OH | |
01,392 | |
15,5241 | |
Solubility in water: 25g/L (20°C). Other solubilities: miscible with alcohol, ether, benzene, oils,, chloroform | |
CC(C)CCO | |
88.15 | |
4.3 mPa.s (20°C) | |
88.15 | |
Biochemistry |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin
GHS Signal Word: Warning
EINECSNumber : 204-633-5
RUO – Research Use Only