missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methyl-1-pentanol 99.0+%, TCI America™
Supplier: TCI America M0600100ML
Specifications
3-Methyl-1-pentanol | |
Yellow | |
C6H14O | |
100 mL | |
3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol | |
CCC(C)CCO | |
102.177 | |
CHEBI:87381 | |
≥99.0% (GC) |
589-35-5 | |
152°C | |
MFCD00002937 | |
1987 | |
IWTBVKIGCDZRPL-UHFFFAOYSA-N | |
3-methylpentan-1-ol | |
11508 | |
102.18 | |
Liquid |
Chemical Identifiers
589-35-5 | |
102.177 | |
IWTBVKIGCDZRPL-UHFFFAOYSA-N | |
11508 | |
3-methylpentan-1-ol |
C6H14O | |
MFCD00002937 | |
3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol | |
CHEBI:87381 | |
CCC(C)CCO |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes