missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1-pentanol 99.0+%, TCI America™

Supplier:  TCI America M060010ML

 View more versions of this product

Catalog No. M060010ML


Explore available promotions

Description

Description

Specifications

Specifications

3-Methyl-1-pentanol
Yellow
C6H14O
10 mL
3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol
CCC(C)CCO
102.177
CHEBI:87381
≥99.0% (GC)
589-35-5
152°C
MFCD00002937
1987
IWTBVKIGCDZRPL-UHFFFAOYSA-N
3-methylpentan-1-ol
11508
102.18
Liquid

Chemical Identifiers

589-35-5
102.177
IWTBVKIGCDZRPL-UHFFFAOYSA-N
11508
3-methylpentan-1-ol
C6H14O
MFCD00002937
3-methyl-1-pentanol, 3-methylpentanol, 1-pentanol, 3-methyl, 2-ethyl-4-butanol, 3-ethyl-1-butanol, 3-ethylbutanol, 3-methyl pentanol, fema no. 3762, 3-methyl pentan-1-ol, 3-methyl-pentan-1-ol
CHEBI:87381
CCC(C)CCO
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions