Learn More
3-Methyl-2,4-pentanedione, mixture of tautomers, 95%, Thermo Scientific Chemicals
$70.48 - $202.71
Chemical Identifiers
CAS | 815-57-6 |
---|---|
Molecular Formula | C6H10O2 |
Molecular Weight (g/mol) | 114.144 |
MDL Number | MFCD00008762 |
InChI Key | GSOHKPVFCOWKPU-UHFFFAOYSA-N |
Synonym | 3-methyl-2,4-pentanedione, methylacetylacetone, 1,1-diacetylethane, 2,4-pentanedione, 3-methyl, methyl acetylacetone, 3-acetylbutan-2-one, 3-methyl-acetylacetone, acmc-209plp, 3-methyl-2,4-pentandione, 3-methylpentan-2,4-dione |
PubChem CID | 69949 |
IUPAC Name | 3-methylpentane-2,4-dione |
SMILES | CC(C(=O)C)C(=O)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0113506
|
Thermo Scientific Chemicals
L0113506 |
5 g |
N/A
|
|
||||||
AAL0113514
|
Thermo Scientific Chemicals
L0113514 |
25 g |
N/A
|
|
||||||
Description
3-Methyl-2,4-pentanedione involves in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands can occur.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methyl-2,4-pentanedione involves in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands can occur.
Solubility
Soluble in water.
Notes
Air & heat sensitive. Do not store together with strong bases and oxidizing agents.
Chemical Identifiers
815-57-6 | |
114.144 | |
GSOHKPVFCOWKPU-UHFFFAOYSA-N | |
69949 | |
CC(C(=O)C)C(=O)C |
C6H10O2 | |
MFCD00008762 | |
3-methyl-2,4-pentanedione, methylacetylacetone, 1,1-diacetylethane, 2,4-pentanedione, 3-methyl, methyl acetylacetone, 3-acetylbutan-2-one, 3-methyl-acetylacetone, acmc-209plp, 3-methyl-2,4-pentandione, 3-methylpentan-2,4-dione | |
3-methylpentane-2,4-dione |
Specifications
815-57-6 | |
170°C to 171°C | |
C6H10O2 | |
MFCD00008762 | |
UN1224 | |
3-methyl-2,4-pentanedione, methylacetylacetone, 1,1-diacetylethane, 2,4-pentanedione, 3-methyl, methyl acetylacetone, 3-acetylbutan-2-one, 3-methyl-acetylacetone, acmc-209plp, 3-methyl-2,4-pentandione, 3-methylpentan-2,4-dione | |
GSOHKPVFCOWKPU-UHFFFAOYSA-N | |
3-methylpentane-2,4-dione | |
69949 | |
95% |
0.98 | |
56°C (132°F) | |
1.442 | |
5 g | |
635909 | |
Soluble in water. | |
CC(C(=O)C)C(=O)C | |
114.144 | |
114.14 | |
3-Methyl-2,4-pentanedione, mixture of tautomers |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber : 212-420-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only