missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 414020500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate | |
97.5 | |
270.0°C to 276.0°C | |
8% max. | |
97.5% min. (HPLC) | |
HCl·H2O | |
50 g | |
2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 3-methyl-2-benzothiazolinone hydrazone hcl monohydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, e-3-methyl-1,3-benzothiazol-2-ylidene hydrazine hydrate hydrochloride, e-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 2z-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrate hydrochloride | |
IYXXQOGEFHAQGU-PIHABLKOSA-N | |
(E)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrate;hydrochloride | |
6508671 | |
98% |
38894-11-0 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
MFCD00149370 | |
(to hydroxyl groups) Passes Test | |
Solubility in water: soluble | |
CN1C2=CC=CC=C2SC1=NN.O.Cl | |
233.72 | |
233.72 | |
Crystalline Powder and/or Chunks |
Chemical Identifiers
38894-11-0 | |
233.72 | |
IYXXQOGEFHAQGU-PIHABLKOSA-N | |
6508671 | |
CN1C2=CC=CC=C2SC1=NN.O.Cl |
HCl·H2O | |
MFCD00149370 | |
2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate, 3-methyl-2-benzothiazolinone hydrazone hcl monohydrate, 3-methyl-2-benzothiazolinone hydrazone hydrochloride hydrate, e-3-methyl-1,3-benzothiazol-2-ylidene hydrazine hydrate hydrochloride, e-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride hydrate, 2z-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrate hydrochloride | |
(E)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrate;hydrochloride |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
GHS Signal Word: Danger
RUO – Research Use Only