Learn More
3-Methyl-2-butenal, 97%, Thermo Scientific Chemicals
$128.56 - $361.62
Chemical Identifiers
CAS | 107-86-8 |
---|---|
Molecular Formula | C5H8O |
Molecular Weight (g/mol) | 84.12 |
InChI Key | SEPQTYODOKLVSB-UHFFFAOYSA-N |
Synonym | 3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde |
PubChem CID | 61020 |
ChEBI | CHEBI:15825 |
IUPAC Name | 3-methylbut-2-enal |
SMILES | CC(=CC=O)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC370180050
|
Thermo Scientific Chemicals
370180050 |
5 mL | Glass bottle |
Each for $128.56
|
|
|||||
AC370180250
|
Thermo Scientific Chemicals
370180250 |
25 mL | Glass bottle |
Each for $361.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-86-8 | |
84.12 | |
3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde | |
CHEBI:15825 | |
CC(=CC=O)C |
C5H8O | |
SEPQTYODOKLVSB-UHFFFAOYSA-N | |
61020 | |
3-methylbut-2-enal |
Specifications
107-86-8 | |
100.0 | |
0.8800g/mL | |
33°C | |
96% min. (GC) | |
C5H8O | |
(CH3)2C=CHCHO | |
01,IV,3464 | |
15,8587 | |
Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether | |
CC(=CC=O)C | |
84.12 | |
CHEBI:15825 | |
97% | |
3-Methyl-2-butenal |
96.0 | |
Colorless to Yellow | |
132.0°C to 133.0°C | |
Authentic | |
Glass bottle | |
1.4610 to 1.4630 | |
5 mL | |
0.88 | |
3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde | |
SEPQTYODOKLVSB-UHFFFAOYSA-N | |
3-methylbut-2-enal | |
61020 | |
84.12 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
Toxic if inhaled.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger
EINECSNumber : 203-527-6
RUO – Research Use Only