missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2-oxobutanoic acid, sodium salt, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 189720010

 View more versions of this product

Catalog No. AC189720010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methyl-2-oxobutanoic acid, sodium salt
3715-29-5
100.0
Beige to Yellow or White
98+%
C5H7NaO3
MFCD00002581
03, 682
WIQBZDCJCRFGKA-UHFFFAOYSA-M
sodium;3-methyl-2-oxobutanoate
2724059
98+%
98+%
98.0
227°C to 231°C
Authentic
Glass bottle
(CH3)2CHCOCO2Na
1 g
sodium 3-methyl-2-oxobutanoate, ketovaline sodium salt, unii-5om3h751iu, butanoic acid, 3-methyl-2-oxo-, sodium salt, 3-methyl-2-oxobutyric acid sodium salt, 2-ketoisovaleric acid sodium salt, sodium 3-methyl-2-oxobutyrate, 3-methyl-2-oxobutanoic acid sodium salt, sodium dimethylpyruvate, ketovaline sodium
CC(C)C(=O)C(=O)[O-].[Na+]
138.1
138.1
Crystalline Powder or Crystals

Chemical Identifiers

3715-29-5
138.1
WIQBZDCJCRFGKA-UHFFFAOYSA-M
2724059
CC(C)C(=O)C(=O)[O-].[Na+]
C5H7NaO3
MFCD00002581
sodium 3-methyl-2-oxobutanoate, ketovaline sodium salt, unii-5om3h751iu, butanoic acid, 3-methyl-2-oxo-, sodium salt, 3-methyl-2-oxobutyric acid sodium salt, 2-ketoisovaleric acid sodium salt, sodium 3-methyl-2-oxobutyrate, 3-methyl-2-oxobutanoic acid sodium salt, sodium dimethylpyruvate, ketovaline sodium
sodium;3-methyl-2-oxobutanoate
Safety and Handling

Safety and Handling

EINECSNumber : 223-062-2

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only